1,3-Dibrom-5-(trifluormethoxy)benzol 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD53962-10GEA 17 EUR
BLDPBD53962-10G BLDPBD53962-500G BLDPBD53962-100G BLDPBD53962-5G BLDPBD53962-1G BLDPBD53962-25G
1,3-Dibrom-5-(trifluormethoxy)benzol 95%
1,3-Dibromo-5-(trifluoromethoxy)benzene

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₃Br₂F₃O
Molecular Weight: 319,90 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00075280
CAS-Nummer: 207226-31-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR