2-Amino-N-methylbenzamid 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD5162-10GEA 30 EUR
BLDPBD5162-10G BLDPBD5162-5G
2-Amino-N-methylbenzamid 98%
2-Amino-N-methylbenzamid
Formel: C₈H₁₀N₂O
Molecular Weight: 150,1778 g/mol
MDL: MFCD00060602
CAS-Nummer: 4141-08-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR