4',5'-Dimethoxy-2'-nitroacetophenone 95%

BLDPBD50763-250MGEA 41 EUR
BLDPBD50763-250MG BLDPBD50763-5G BLDPBD50763-1G
4',5'-Dimethoxy-2'-nitroacetophenone 95%
4',5'-Dimethoxy-2'-nitroacetophenone

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₁₁NO₅
Molecular Weight: 225,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00130048
CAS-Nummer: 4101-32-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR