3-(Methylsulfonyl)phenylboronsäure 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD5034-100MGEA 6 EUR
BLDPBD5034-100MG BLDPBD5034-25G BLDPBD5034-10G BLDPBD5034-1G BLDPBD5034-5G BLDPBD5034-250MG
3-(Methylsulfonyl)phenylboronsäure 98%
3-(Methylsulfonyl)phenylboronsäure

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₉BO₄S
Molecular Weight: 200,02 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03092935
CAS-Nummer: 373384-18-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR