1-(2-Bromethoxy)-3-methoxybenzol 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD49867-25GEA 288 EUR
BLDPBD49867-25G BLDPBD49867-5G BLDPBD49867-1G BLDPBD49867-250MG
1-(2-Bromethoxy)-3-methoxybenzol 98%
1-(2-Bromoethoxy)-3-methoxybenzene

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₁₁O₂Br
Molecular Weight: 231,0864 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00980056
CAS-Nummer: 3245-45-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR