4-Methyl-3-nitrophenylboronsäure 98%

BLDPBD4917-1GEA 9 EUR
BLDPBD4917-1G BLDPBD4917-25G BLDPBD4917-10G BLDPBD4917-5G
4-Methyl-3-nitrophenylboronsäure 98%
4-Methyl-3-nitrophenylboronsäure
Formel: C₇H₈BNO₄
Molecular Weight: 180,96 g/mol
MDL: MFCD00191550
CAS-Nummer: 80500-27-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR