9-(3-Biphenylyl)-3-bromocarbazole 97%

BLDPBD491114-1GEA 10 EUR
BLDPBD491114-1G BLDPBD491114-5G BLDPBD491114-25G
9-(3-Biphenylyl)-3-bromocarbazole 97%
9-(3-Biphenylyl)-3-bromocarbazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₂₄H₁₆BrN
Molecular Weight: 398,30 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28138081
CAS-Nummer: 1428551-28-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR