5-Bromo-7H-benzo[c]carbazole 98%
|
|
BLDPBD491098-25GEA
477
EUR
BLDPBD491098-25G
BLDPBD491098-5G
BLDPBD491098-1G
BLDPBD491098-250MG
BLDPBD491098-0.025
5-Bromo-7H-benzo[c]carbazole 98%
5-Bromo-7H-benzo[c]carbazole
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formel:
C₁₆H₁₀BrN Molecular Weight: 296,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30063204 CAS-Nummer: 131409-18-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...