3-Phenyl-9H-carbazole 98%

BLDPBD491004-1GEA 7 EUR
BLDPBD491004-1G BLDPBD491004-250MG BLDPBD491004-100G BLDPBD491004-10G BLDPBD491004-25G BLDPBD491004-5G BLDPBD491004-500G
3-Phenyl-9H-carbazole 98%
3-Phenyl-9H-carbazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₈H₁₃N
Molecular Weight: 243,31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22052094
CAS-Nummer: 103012-26-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR