3-Fluor-4-methylphenylboronsäure 97%
|
|
BLDPBD4899-10GEA
29
EUR
BLDPBD4899-10G
BLDPBD4899-1G
BLDPBD4899-25G
BLDPBD4899-5G
3-Fluor-4-methylphenylboronsäure 97%
3-Fluor-4-methylphenylboronsäure
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formel:
C₇H₈BFO₂ Molecular Weight: 153,95 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02683115 CAS-Nummer: 168267-99-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...