3-Chlor-4-methylphenylboronsäure 97%

BLDPBD4891-10GEA 53 EUR
BLDPBD4891-10G BLDPBD4891-5G BLDPBD4891-100G BLDPBD4891-25G BLDPBD4891-1G
3-Chlor-4-methylphenylboronsäure 97%
3-Chlor-4-methylphenylboronsäure

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₈BClO₂
Molecular Weight: 170,40 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04039010
CAS-Nummer: 175883-63-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR