3-Hydroxybenzamide 95%

BLDPBD4836-250MGEA 22 EUR
BLDPBD4836-250MG BLDPBD4836-1G BLDPBD4836-100MG BLDPBD4836-5G BLDPBD4836-10G
3-Hydroxybenzamide 95%
3-Hydroxybenzamide

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₇NO₂
Molecular Weight: 137,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00017572
CAS-Nummer: 618-49-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR