2-Hydrazinobenzothiazol 95%

BLDPBD4825-25GEA 122 EUR
BLDPBD4825-25G BLDPBD4825-1G
2-Hydrazinobenzothiazol 95%
2-Hydrazinobenzothiazole

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formel: C₇H₇N₃S
Molecular Weight: 165,21 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00041849
CAS-Nummer: 615-21-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR