3,5-Dibrom-p-toluidin 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD4731-250MGEA 26 EUR
BLDPBD4731-250MG BLDPBD4731-5G BLDPBD4731-1G BLDPBD4731-25G
3,5-Dibrom-p-toluidin 98%
3,5-Dibrom-p-toluidin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₇Br₂N
Molecular Weight: 264,96 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00151806
CAS-Nummer: 13194-73-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR