(4-Bromphenyl)methansulfonsäurechlorid 98%

BLDPBD4506-250MGEA 33 EUR
BLDPBD4506-250MG BLDPBD4506-1G BLDPBD4506-5G
(4-Bromphenyl)methansulfonsäurechlorid 98%
(4-Bromophenyl)methanesulphonyl chloride

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₆BrClO₂S
Molecular Weight: 269,5433 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04973454
CAS-Nummer: 53531-69-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR