Methyl-(3-bromphenyl)acetat 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD45056-500GEA 1130 EUR
BLDPBD45056-500G BLDPBD45056-10G BLDPBD45056-1G BLDPBD45056-5G BLDPBD45056-25G BLDPBD45056-100G
Methyl-(3-bromphenyl)acetat 98%
Methyl-(3-bromphenyl)acetat

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₉BrO₂
Molecular Weight: 229,0700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06858764
CAS-Nummer: 150529-73-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR