3-Carboxy-5-nitrophenylboronsäure 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD4487-5GEA 33 EUR
BLDPBD4487-5G BLDPBD4487-250MG BLDPBD4487-1G BLDPBD4487-10G
3-Carboxy-5-nitrophenylboronsäure 97%
3-Carboxy-5-nitrophenylboronsäure

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₆BNO₆
Molecular Weight: 210,94 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00757433
CAS-Nummer: 101084-81-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR