3-Carboxy-4-fluorphenylboronsäure 98%
Lieferant: BLD PHARMATECH GMBH
|
|
BLDPBD4480-1GEA
11
EUR
BLDPBD4480-1G
BLDPBD4480-25G
BLDPBD4480-5G
3-Carboxy-4-fluorphenylboronsäure 98%
3-Carboxy-4-fluorphenylboronsäure
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₇H₆BFO₄ Molecular Weight: 183,93 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095129 CAS-Nummer: 872460-12-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...