1,3-Benzothiazol-2(3H)-on 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD4470-10GEA 21 EUR
BLDPBD4470-10G BLDPBD4470-100G BLDPBD4470-25G BLDPBD4470-5G
1,3-Benzothiazol-2(3H)-on 98%
Benzothiazol-2-ol

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₅NOS
Molecular Weight: 151,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00792539
CAS-Nummer: 934-34-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR