4-Fluorsalicylaldehyd 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD4425-5GEA 78 EUR
BLDPBD4425-5G BLDPBD4425-250MG BLDPBD4425-100G BLDPBD4425-25G BLDPBD4425-10G BLDPBD4425-1G
4-Fluorsalicylaldehyd 97%
4-Fluorsalicylaldehyd

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₅FO₂
Molecular Weight: 140,1118 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03788526
CAS-Nummer: 348-28-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR