(2-Carboxyethyl)triphenylphosphoniumbromid 97%

BLDPBD44166-100GEA 121 EUR
BLDPBD44166-100G BLDPBD44166-1G BLDPBD44166-0.025G BLDPBD44166-25G BLDPBD44166-500G BLDPBD44166-5G BLDPBD44166-10G
(2-Carboxyethyl)triphenylphosphoniumbromid 97%
(2-Carboxyethyl)triphenylphosphonium bromide

Storage condition: Inert atmosphere, Room Temperature

Formel: C₂₁H₂₀BrO₂P
Molecular Weight: 415,2601 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00031698
CAS-Nummer: 51114-94-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR