2-Chlor-4-fluor-5-nitroanisol 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD4361-1GEA 16 EUR
BLDPBD4361-1G BLDPBD4361-100MG BLDPBD4361-25G BLDPBD4361-5G BLDPBD4361-250MG
2-Chlor-4-fluor-5-nitroanisol 97%
2-Chlor-4-fluor-5-nitroanisol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₅ClFNO₃
Molecular Weight: 205,5700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03425810
CAS-Nummer: 84478-76-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR