2-Amino-5-pyrimidinboronsäurepinakolester 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD42692-1GEA 13 EUR
BLDPBD42692-1G BLDPBD42692-10G BLDPBD42692-5G BLDPBD42692-25G BLDPBD42692-100G
2-Amino-5-pyrimidinboronsäurepinakolester 98%
2-Amino-5-pyrimidinboronsäurepinakolester

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₀H₁₆BN₃O₂
Molecular Weight: 221,06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06795685
CAS-Nummer: 402960-38-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR