4-(Trifluormethyl)nicotinsäure 98%
|
|
BLDPBD4230-10GEA
37
EUR
BLDPBD4230-0.025G
BLDPBD4230-10G
BLDPBD4230-250MG
BLDPBD4230-25G
BLDPBD4230-1G
BLDPBD4230-100G
BLDPBD4230-5G
4-(Trifluormethyl)nicotinsäure 98%
4-(Trifluormethyl)nicotinsäure
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₇H₄F₃NO₂ Molecular Weight: 191,11 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082626 CAS-Nummer: 158063-66-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...