4'-Demethylepipodophyllotoxin 98%

BLDPBD41925-250MGEA 13 EUR
BLDPBD41925-250MG BLDPBD41925-25G BLDPBD41925-1G
4'-Demethylepipodophyllotoxin 98%
4'-Demethylepipodophyllotoxin

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₂₁H₂₀O₈
Molecular Weight: 400,38 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00189421
CAS-Nummer: 6559-91-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR