Histamin (freie Base) 97%

BLDPBD41747-100MGEA 10 EUR
BLDPBD41747-100MG
Histamin (freie Base) 97%
Histamin (freie Base)

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formel: C₅H₉N₃
Molecular Weight: 111,15 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00005210
CAS-Nummer: 51-45-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR