3-Bromo-5-iodobenzaldehyde 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD4135-100MGEA 51 EUR
BLDPBD4135-100MG BLDPBD4135-250MG BLDPBD4135-1G
3-Bromo-5-iodobenzaldehyde 95%
3-Bromo-5-iodobenzaldehyde

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₇H₄BrIO
Molecular Weight: 310,915 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07779024
CAS-Nummer: 188813-09-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR