2-Brom-4-iodbenzaldehyd 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD4134-25GEA 1347 EUR
BLDPBD4134-25G BLDPBD4134-100MG BLDPBD4134-1G BLDPBD4134-5G BLDPBD4134-250MG
2-Brom-4-iodbenzaldehyd 97%
2-Bromo-4-iodobenzaldehyde

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₇H₄BrIO
Molecular Weight: 310,9200 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07779025
CAS-Nummer: 261903-03-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR