6-Methyl-3(2H)-pyridazinon 97%
Lieferant: BLD PHARMATECH GMBH
|
|
BLDPBD41253-1GEA
6
EUR
BLDPBD41253-1G
BLDPBD41253-10G
BLDPBD41253-25G
BLDPBD41253-5G
BLDPBD41253-100G
6-Methyl-3(2H)-pyridazinon 97%
6-Methyl-3(2H)-pyridazinon
Storage condition: Sealed in dry, Room Temperature
Formel:
C₅H₆N₂O Molecular Weight: 110,12 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039720 CAS-Nummer: 13327-27-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...