1,8-Dibromonaphthalene 98%
|
|
BLDPBD41002-500GEA
1955
EUR
BLDPBD41002-500G
BLDPBD41002-250MG
BLDPBD41002-10G
BLDPBD41002-100G
BLDPBD41002-25G
BLDPBD41002-5G
BLDPBD41002-1G
1,8-Dibromonaphthalene 98%
1,8-Dibromonaphthalene
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formel:
C₁₀H₆Br₂ Molecular Weight: 285,97 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00183574 CAS-Nummer: 17135-74-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...