1-Brom-3,5-di-tert-butylbenzol 98%

BLDPBD41000-1GEA 6 EUR
BLDPBD41000-1G BLDPBD41000-25G BLDPBD41000-10G BLDPBD41000-500G BLDPBD41000-5G BLDPBD41000-100G
1-Brom-3,5-di-tert-butylbenzol 98%
1-Brom-3,5-di-tert-butylbenzol

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₄H₂₁Br
Molecular Weight: 269,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00796945
CAS-Nummer: 22385-77-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR