Ethyl-3-amino-1H-pyrazol-4-carboxylat 95%
|
|
BLDPBD40841-1GEA
6
EUR
BLDPBD40841-1G
BLDPBD40841-500G
BLDPBD40841-100G
BLDPBD40841-25G
BLDPBD40841-5G
BLDPBD40841-10G
Ethyl-3-amino-1H-pyrazol-4-carboxylat 95%
Ethyl-3-amino-1H-pyrazol-4-carboxylat
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formel:
C₆H₉N₃O₂ Molecular Weight: 155,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005238 CAS-Nummer: 6994-25-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...