Tetrabutylammoniumfluorid Hydrat 85%
|
|
BLDPBD40693-25GEA
7
EUR
BLDPBD40693-25G
Tetrabutylammoniumfluorid Hydrat 85%
Tetrabutylammoniumfluorid Hydrat
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₁₆H₃₆FN·H₂O Molecular Weight: 279,4774 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149980 CAS-Nummer: 22206-57-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...