3,5,6-Trichlorsalicylsäure 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD4048-25GEA 11 EUR
BLDPBD4048-25G BLDPBD4048-100G BLDPBD4048-500G
3,5,6-Trichlorsalicylsäure 98%
3,5,6-Trichlorsalicylsäure

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₃Cl₃O₃
Molecular Weight: 241,46 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00075245
CAS-Nummer: 40932-60-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR