4-Methoxy-2-(trifluormethoxy)benzonitril 98%

BLDPBD399362-250MGEA 51 EUR
BLDPBD399362-250MG BLDPBD399362-5G BLDPBD399362-1G BLDPBD399362-100MG
4-Methoxy-2-(trifluormethoxy)benzonitril 98%
4-Methoxy-2-(trifluormethoxy)benzonitril

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₆F₃NO₂
Molecular Weight: 217,1447 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06660284
CAS-Nummer: 886502-33-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR