3-Amino-5-tert-butyl-1H-pyrazol 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD3885-25GEA 10 EUR
BLDPBD3885-25G BLDPBD3885-10G BLDPBD3885-100G
3-Amino-5-tert-butyl-1H-pyrazol 97%
3-Amino-5-tert-butyl-1H-pyrazol

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₁₃N₃
Molecular Weight: 139,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00067987
CAS-Nummer: 82560-12-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR