Ethyl-5-amino-1-methyl-1H-pyrazol-4-carboxylat 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD3838-1000GEA 370 EUR
BLDPBD3838-1000G BLDPBD3838-100G BLDPBD3838-500G BLDPBD3838-25G BLDPBD3838-10G BLDPBD3838-5G
Ethyl-5-amino-1-methyl-1H-pyrazol-4-carboxylat 97%
Ethyl-5-amino-1-methyl-1H-pyrazol-4-carboxylat

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₁₁N₃O₂
Molecular Weight: 169,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00051652
CAS-Nummer: 31037-02-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR