4-(Chlormethyl)-3,5-dimethylisoxazol 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD3683-1GEA 7 EUR
BLDPBD3683-1G BLDPBD3683-25G BLDPBD3683-5G BLDPBD3683-10G
4-(Chlormethyl)-3,5-dimethylisoxazol 97%
4-(Chloromethyl)-3,5-dimethylisoxazole

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₆H₈ClNO
Molecular Weight: 145,59 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00003154
CAS-Nummer: 19788-37-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR