Sulfachinoxalin Natriumsalz 98%
|
|
BLDPBD35528-1GEA
94
EUR
BLDPBD35528-1G
BLDPBD35528-250MG
BLDPBD35528-100MG
Sulfachinoxalin Natriumsalz 98%
Sulfachinoxalin Natriumsalz
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₁₄H₁₁N₄NaO₂S Molecular Weight: 322,3175 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00799608 CAS-Nummer: 967-80-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...