4-Nitro-2-picolin-N-oxid 98%

Lieferant: BLD PHARMATECH GMBH

Synonyme: 2-Methyl-4-nitropyridin-1-oxid2-Methyl-4-nitropyridin-N-oxid4-Nitro-2-picolin-N-oxid

BLDPBD3527-1GEA 7 EUR
BLDPBD3527-1G BLDPBD3527-5G BLDPBD3527-10G BLDPBD3527-25G
4-Nitro-2-picolin-N-oxid 98%
4-Nitro-2-picolin-N-oxid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₆H₆N₂O₃
Molecular Weight: 154,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00014625
CAS-Nummer: 5470-66-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR