4-Brom-2,5-xylenol 97%
|
|
BLDPBD35122-25GEA
481
EUR
BLDPBD35122-25G
BLDPBD35122-1G
BLDPBD35122-250MG
BLDPBD35122-100G
BLDPBD35122-5G
BLDPBD35122-10G
4-Brom-2,5-xylenol 97%
4-Brom-2,5-xylenol
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₈H₉BrO Molecular Weight: 201,0600 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00029746 CAS-Nummer: 85223-93-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...