3,5-Dimethyl-4-ethoxyphenylboronsäure 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD35110-25GEA 161 EUR
BLDPBD35110-25G BLDPBD35110-1G BLDPBD35110-0.025G BLDPBD35110-100G BLDPBD35110-5G
3,5-Dimethyl-4-ethoxyphenylboronsäure 98%
3,5-Dimethyl-4-ethoxyphenylboronsäure

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₀H₁₅BO₃
Molecular Weight: 194,04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02179463
CAS-Nummer: 850568-59-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR