6-Indolboronsäurepinakolester 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD34486-25GEA 822 EUR
BLDPBD34486-25G BLDPBD34486-5G BLDPBD34486-1G BLDPBD34486-100MG BLDPBD34486-250MG
6-Indolboronsäurepinakolester 97%
6-Indolboronsäurepinakolester

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₄H₁₈BNO₂
Molecular Weight: 243,1092 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07368852
CAS-Nummer: 642494-36-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR