3-Methyl-2-thiophencarbonitril 98%
|
|
BLDPBD3438-10GEA
105
EUR
BLDPBD3438-10G
BLDPBD3438-1G
BLDPBD3438-25G
BLDPBD3438-5G
3-Methyl-2-thiophencarbonitril 98%
3-Methyl-2-thiophencarbonitril
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formel:
C₆H₅NS Molecular Weight: 123,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052086 CAS-Nummer: 55406-13-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...