Methyl 5-Methoxy-2-thiophenecarboxylate 95+%

Lieferant: BLD PHARMATECH GMBH

BLDPBD338400-100MGEA 33 EUR
BLDPBD338400-100MG BLDPBD338400-250MG BLDPBD338400-1G BLDPBD338400-5G
Methyl 5-Methoxy-2-thiophenecarboxylate 95+%
Methyl 5-Methoxy-2-thiophenecarboxylate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₈O₃S
Molecular Weight: 172,202 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22682797
CAS-Nummer: 77133-25-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR