2-Bromo-6-iodoaniline 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD335475-100MGEA 38 EUR
BLDPBD335475-100MG BLDPBD335475-1G BLDPBD335475-250MG
2-Bromo-6-iodoaniline 97%
2-Bromo-6-iodoaniline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₆H₅BrIN
Molecular Weight: 297,919 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18391615
CAS-Nummer: 84483-27-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR