9-Chloro-2,3-dihydro-1H-cyclopenta[b]quinoline 97%

BLDPBD334306-250MGEA 270 EUR
BLDPBD334306-250MG BLDPBD334306-100MG BLDPBD334306-1G
9-Chloro-2,3-dihydro-1H-cyclopenta[b]quinoline 97%
9-Chloro-2,3-dihydro-1H-cyclopenta[b]quinoline

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₂H₁₀ClN
Molecular Weight: 203,668 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD05670710
CAS-Nummer: 40528-00-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR