4-Aminotetrahydropyran Hydrochlorid 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD33238-10GEA 22 EUR
BLDPBD33238-10G BLDPBD33238-100G BLDPBD33238-1G BLDPBD33238-5G BLDPBD33238-25G
4-Aminotetrahydropyran Hydrochlorid 97%
4-Aminotetrahydropyran Hydrochlorid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₅H₁₂ClNO
Molecular Weight: 137,6079 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00100881
CAS-Nummer: 33024-60-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR