2,3-Furandicarbonsäure 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD3300-250MGEA 29 EUR
BLDPBD3300-250MG BLDPBD3300-100MG BLDPBD3300-1G BLDPBD3300-5G
2,3-Furandicarbonsäure 95%
Furan-2,3-dicarboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₆H₄O₅
Molecular Weight: 156,0930 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01860231
CAS-Nummer: 4282-24-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR