4-Bromo-5-chlorobenzene-1,2-diamine 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD328456-250MGEA 53 EUR
BLDPBD328456-250MG BLDPBD328456-1G BLDPBD328456-100MG
4-Bromo-5-chlorobenzene-1,2-diamine 95%
4-Bromo-5-chlorobenzene-1,2-diamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₆H₆BrClN₂
Molecular Weight: 221,482 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16987754
CAS-Nummer: 75293-95-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR